CH6371378377 Chem Impex International, Inc.

Nalpha,Nomega,Nomega'-Tris-Z-D-arginine

  • MDL number MFCD00237330

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-139 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form Yellowish powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-n-i-sup-omega-sup-i-n-i-sup-omega-sup-tris-z-d-arginine/2574?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-n-i-sup-omega-sup-i-n-i-sup-omega-sup-tris-z-d-arginine/2574?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart