CH6371378378 Chem Impex International, Inc.

Na-Z-Nw-(4-methoxy-2,3,6-trimethylbenzenesulfonyl)-D-arginine cyclohexylammonium salt

  • MDL number MFCD00153321
  • Cas Number : 210557-94-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 196-199
Synonym Z-D-Arg(Mtr)-OH CHA
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-z-i-n-i-sup-sup-4-methoxy-2-3-6-trimethylbenzenesulfonyl-d-arginine-cyclohexylammonium-salt/2575?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-z-i-n-i-sup-sup-4-methoxy-2-3-6-trimethylbenzenesulfonyl-d-arginine-cyclohexylammonium-salt/2575?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart