CH6371378381 Chem Impex International, Inc.

Z-L-aspartic acid b-tert-butyl ester monohydrate

  • MDL number MFCD00150667
  • Cas Number : 5545-52-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-L-Asp(OtBu)-OH H2O
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-aspartic-acid-ß-i-tert-i-butyl-ester-monohydrate/2578?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-aspartic-acid-ß-i-tert-i-butyl-ester-monohydrate/2578?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart