CH6371378383 Chem Impex International, Inc.

Z-D-aspartic acid b-tert-butyl ester hydrate

  • MDL number MFCD00153327
  • Cas Number : 71449-08-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50-61
Synonym Z-D-Asp(OtBu)-OH H2O
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-d-aspartic-acid-beta-i-tert-i-butyl-ester-hydrate/2580?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-d-aspartic-acid-beta-i-tert-i-butyl-ester-hydrate/2580?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart