CH6371378384 Chem Impex International, Inc.

Z-D-aspartic acid b-benzyl ester

  • MDL number MFCD00237336
  • Cas Number : 5241-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-108
Synonym Z-D-Asp(OBzl)-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-d-aspartic-acid-ß-benzyl-ester/2581?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-d-aspartic-acid-ß-benzyl-ester/2581?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart