CH6371378385 Chem Impex International, Inc.

Z-L-glutamic acid g-tert-butyl ester a-N-hydroxysuccinimide ester

  • MDL number MFCD00153329
  • Cas Number : 4666-16-4
  • Molecular Weight : 434.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104-112° C
Purity 97
Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-glutamic-acid-g-i-tert-i-butyl-ester-a-i-n-i-hydroxysuccinimide-ester/2582?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-glutamic-acid-g-i-tert-i-butyl-ester-a-i-n-i-hydroxysuccinimide-ester/2582?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart