CH6371378395 Chem Impex International, Inc.

Z-L-proline N-hydroxysuccinimide ester

  • MDL number MFCD00042757
  • Cas Number : 3397-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88 - 92 °C
Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-proline-i-n-i-hydroxysuccinimide-ester/2592?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-proline-i-n-i-hydroxysuccinimide-ester/2592?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart