CH6371378397 Chem Impex International, Inc.

Z-L-threonine N-hydroxysuccinimide ester

  • MDL number MFCD00153344
  • Cas Number : 76401-90-6
  • Molecular Weight : 350.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97-101 °C
Purity 90
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-threonine-i-n-i-hydroxysuccinimide-ester/2594?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-threonine-i-n-i-hydroxysuccinimide-ester/2594?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart