CH6371378398 Chem Impex International, Inc.

Z-O-tert-butyl-D-threonine dicyclohexylammonium salt

  • MDL number MFCD00153345
  • Cas Number : 201275-65-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128-134 °C
Purity 99
Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-i-o-i-i-tert-i-butyl-d-threonine-dicyclohexylammonium-salt/2596?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-i-o-i-i-tert-i-butyl-d-threonine-dicyclohexylammonium-salt/2596?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart