CH6371378401 Chem Impex International, Inc.

Z-O-benzyl-D-tyrosine

  • MDL number MFCD00153349
  • Cas Number : 92455-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113-119� C
Synonym Z-D-Tyr(Bzl)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-i-o-i-benzyl-d-tyrosine/2600?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-i-o-i-benzyl-d-tyrosine/2600?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart