CH6371378404 Chem Impex International, Inc.

Z-L-phenylglycine N-hydroxysuccinimide ester

  • MDL number MFCD00153339
  • Cas Number : 146118-22-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156-162
Synonym Z-L-Phg-OSu
Purity 98
Storage temp. Store at = -4
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-phenylglycine-i-n-i-hydroxysuccinimide-ester/2603?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-phenylglycine-i-n-i-hydroxysuccinimide-ester/2603?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart