CH6371378405 Chem Impex International, Inc.

Z-L-serine hydrazide

  • MDL number MFCD00153341
  • Cas Number : 26582-86-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182-186� C
Synonym Z-L-Ser-NHNH2
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-serine-hydrazide/2604?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-serine-hydrazide/2604?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart