CH6371378406 Chem Impex International, Inc.

Z-L-threonine hydrazide

  • MDL number MFCD00153343
  • Cas Number : 49706-30-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 193-200
Synonym Z-L-Thr-NHNH2
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-threonine-hydrazide/2605?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-threonine-hydrazide/2605?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart