CH6371378417 Chem Impex International, Inc.

Z-L-norleucine N-hydroxysuccinimide ester

  • MDL number MFCD00237396
  • Cas Number : 36360-62-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77-85
Synonym Z-L-norleucine N-hydroxysuccinimide ester
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-norleucine-i-n-i-hydroxysuccinimide-ester/2618?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-norleucine-i-n-i-hydroxysuccinimide-ester/2618?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart