CH6371378418 Chem Impex International, Inc.

Z-L-norvaline N-hydroxysuccinimide ester

  • MDL number MFCD00237397
  • Cas Number : 71447-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94 - 98
Synonym Z-L-Nva-OSu
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-norvaline-i-n-i-hydroxysuccinimide-ester/2619?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-norvaline-i-n-i-hydroxysuccinimide-ester/2619?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart