CH6371378419 Chem Impex International, Inc.

Nalpha,delta-Bis-Z-L-ornithine N-hydroxysuccinimide ester

  • MDL number MFCD00237359
  • Cas Number : 90970-61-9
  • Molecular Weight : 497.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109-119 °C
Purity 98
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-delta-sup-i-i-bis-i-i-z-l-ornithine-i-n-i-hydroxysuccinimide-ester/2620?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-delta-sup-i-i-bis-i-i-z-l-ornithine-i-n-i-hydroxysuccinimide-ester/2620?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart