CH6371378423 Chem Impex International, Inc.

Nalpha-Z-L-tryptophan N-hydroxysuccinimide ester

  • MDL number MFCD00153347
  • Cas Number : 50305-28-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Physical Form White to cream powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-z-l-tryptophan-i-n-i-hydroxysuccinimide-ester/2624?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-z-l-tryptophan-i-n-i-hydroxysuccinimide-ester/2624?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart