CH6371378427 Chem Impex International, Inc.

Na-Fmoc-Ng-4-methyltrityl-L-asparagine

  • MDL number MFCD00237160
  • Cas Number : 144317-22-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 192-196
Synonym Fmoc-L-Asn(Mtt)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-fmoc-i-n-i-sup-gamma-sup-4-methyltrityl-l-asparagine/2634?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-fmoc-i-n-i-sup-gamma-sup-4-methyltrityl-l-asparagine/2634?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart