CH6371378432 Chem Impex International, Inc.

Fmoc-3,5-dibromo-L-tyrosine

  • MDL number MFCD00065686
  • Cas Number : 201484-26-6
  • Molecular Weight : 561.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184-193 °C
Purity 99
Storage temp. Store at 0-8°C
Physical Form White cyrstals
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-3-5-dibromo-l-tyrosine/2640?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-3-5-dibromo-l-tyrosine/2640?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart