CH6371378438 Chem Impex International, Inc.

Na-Fmoc-Nd-methyltrityl-L-glutamine

  • MDL number MFCD00237163
  • Cas Number : 144317-23-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-fmoc-i-n-i-sup-d-sup-methyltrityl-l-glutamine/2646?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-fmoc-i-n-i-sup-d-sup-methyltrityl-l-glutamine/2646?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart