CH6371378446 Chem Impex International, Inc.

Boc-alpha-aminoisobutyric acid N-hydroxysuccinimide ester

  • MDL number MFCD00672711
  • Cas Number : 104055-39-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162-168 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-alpha-aminoisobutyric-acid-i-n-i-hydroxysuccinimide-ester/2663?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-alpha-aminoisobutyric-acid-i-n-i-hydroxysuccinimide-ester/2663?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart