CH6371378449 Chem Impex International, Inc.

Boc-D-norleucine

  • MDL number MFCD00063168
  • Cas Number : 55674-63-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-D-Nle-OH
Purity 98
Storage temp. Store at 0-8
Physical Form Syrup
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-d-norleucine/2667?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-d-norleucine/2667?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart