CH6371378452 Chem Impex International, Inc.

Boc-4-(Fmoc-amino)-D-phenylalanine

  • MDL number MFCD00151888
  • Cas Number : 173054-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-D-Phe(4-NH-Fmoc)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-4-fmoc-amino-d-phenylalanine/2670?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-4-fmoc-amino-d-phenylalanine/2670?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart