CH6371378453 Chem Impex International, Inc.

Boc-L-Phe(4-NHZ)-OH

  • MDL number MFCD00038751
  • Cas Number : 55533-25-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-4-(Z-amino)-L-phenylalanine
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-phe-4-nhz-oh/2671?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-phe-4-nhz-oh/2671?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart