CH6371378456 Chem Impex International, Inc.

Na-Boc-Nw,Nw'-bis-Z-L-arginine N-hydroxysuccinimide ester

  • MDL number MFCD00236826
  • Cas Number : 76052-29-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-L-Arg(Z)2-OSu
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-fmoc-i-n-i-sup-omega-sup-i-n-i-sup-omega-sup-i-i-bis-i-i-z-l-arginine-i-n-i-hydroxysuccinimide-ester/2674?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-fmoc-i-n-i-sup-omega-sup-i-n-i-sup-omega-sup-i-i-bis-i-i-z-l-arginine-i-n-i-hydroxysuccinimide-ester/2674?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart