CH6371378457 Chem Impex International, Inc.

Boc-L-aspartic acid alpha-9-fluorenylmethyl ester

  • MDL number MFCD00151868
  • Cas Number : 129046-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-aspartic-acid-a-9-fluorenylmethyl-ester/2675?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-aspartic-acid-a-9-fluorenylmethyl-ester/2675?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart