CH6371378458 Chem Impex International, Inc.

Boc-D-aspartic acid b-tert-butyl ester dicyclohexylammonium salt

  • MDL number MFCD00076911
  • Cas Number : 200334-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138-144
Synonym Boc-D-Asp(OtBu)-OH DCHA
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-d-aspartic-acid-beta-i-tert-i-butyl-ester-dicyclohexylammonium-salt/2676?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-d-aspartic-acid-beta-i-tert-i-butyl-ester-dicyclohexylammonium-salt/2676?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart