CH6371378463 Chem Impex International, Inc.

Boc-L-citrulline 4-nitrophenyl ester

  • MDL number MFCD00076998
  • Cas Number : 56612-88-5
  • Molecular Weight : 396.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93-97 °C
Purity 98
Storage temp. Store at 0-8°C
Physical Form Yellow to brown powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-citrulline-4-nitrophenyl-ester/2683?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-citrulline-4-nitrophenyl-ester/2683?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart