CH6371378465 Chem Impex International, Inc.

Boc-b-cyclohexyl-D-alanine dicyclohexylammonium salt

  • MDL number MFCD00155637
  • Cas Number : 198470-07-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 168-173 �C
Synonym Boc-D-Cha-OH DCHA
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-beta-cyclohexyl-d-alanine-dicyclohexylammonium-salt/2685?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-beta-cyclohexyl-d-alanine-dicyclohexylammonium-salt/2685?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart