CH6371378466 Chem Impex International, Inc.

Boc-S-acetamidomethyl-L-cysteine 4-nitrophenyl ester

  • MDL number MFCD00236831
  • Cas Number : 58651-76-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125-133
Synonym Boc-L-Cys(Acm)-ONp
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-acetamidomethyl-l-cysteine-4-nitrophenyl-ester/2686?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-acetamidomethyl-l-cysteine-4-nitrophenyl-ester/2686?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart