CH6371378468 Chem Impex International, Inc.

Boc-S-4-methylbenzyl-L-cysteine N-hydroxysuccinimide ester

  • MDL number MFCD00236833
  • Cas Number : 140842-91-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90-97 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-4-methylbenzyl-l-cysteine-i-n-i-hydroxysuccinimide-ester/2690?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-4-methylbenzyl-l-cysteine-i-n-i-hydroxysuccinimide-ester/2690?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart