CH6371378469 Chem Impex International, Inc.

Boc-S-4-methylbenzyl-D-cysteine

  • MDL number MFCD00076924
  • Cas Number : 61925-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78-85
Synonym Boc-D-Cys(pMeBzl)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-4-methylbenzyl-d-cysteine/2691?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-4-methylbenzyl-d-cysteine/2691?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart