CH6371378470 Chem Impex International, Inc.

Boc-3,5-dibromo-L-tyrosine

  • MDL number MFCD00076993
  • Cas Number : 58960-71-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145-150
Synonym Boc-3,5-dibromo-L-Tyr-OH
Purity 0.99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-3-5-dibromo-l-tyrosine/2693?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-3-5-dibromo-l-tyrosine/2693?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart