CH6371378473 Chem Impex International, Inc.

Boc-S-acetamidomethyl-L-penicillamine

  • MDL number MFCD00151885
  • Cas Number : 129972-45-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-L-Pen(Acm)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-acetamidomethyl-l-penicillamine/2696?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-acetamidomethyl-l-penicillamine/2696?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart