CH6371378474 Chem Impex International, Inc.

Boc-S-acetamidomethyl-D-penicillamine

  • MDL number MFCD00151884
  • Cas Number : 201421-14-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-D-Pen(Acm)-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-acetamidomethyl-d-penicillamine/2697?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-acetamidomethyl-d-penicillamine/2697?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart