CH6371378475 Chem Impex International, Inc.

Boc-S-trityl-L-penicillamine

  • MDL number MFCD00237381
  • Cas Number : 135592-13-1
  • Molecular Weight : 491.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131-137 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-trityl-l-penicillamine/2698?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-trityl-l-penicillamine/2698?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart