CH6371378477 Chem Impex International, Inc.

Boc-L-glutamic acid gamma-benzyl ester alpha-4-nitrophenyl ester

  • MDL number MFCD00038124
  • Cas Number : 7536-59-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115 - 121 °C
Purity 98
Storage temp. Store at 0 - 8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-glutamic-acid-gamma-benzyl-ester-alpha-4-nitrophenyl-ester/2702?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-glutamic-acid-gamma-benzyl-ester-alpha-4-nitrophenyl-ester/2702?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart