CH6371378484 Chem Impex International, Inc.

Boc-L-leucine 4-nitrophenyl ester

  • MDL number MFCD00038291
  • Cas Number : 3350-19-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71-77° C
Purity 97
Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-leucine-4-nitrophenyl-ester/2709?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-leucine-4-nitrophenyl-ester/2709?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart