CH6371378487 Chem Impex International, Inc.

Na-Boc-Ne-2-chloro-Z-L-lysine N-hydroxysuccinimide ester

  • MDL number MFCD00236855
  • Cas Number : 66438-39-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97-104
Synonym Boc-L-Lys(2-Cl-Z)-OSu
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-boc-i-n-i-sup-epsilon-sup-2-chloro-z-l-lysine-i-n-i-hydroxysuccinimide-ester/2712?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-boc-i-n-i-sup-epsilon-sup-2-chloro-z-l-lysine-i-n-i-hydroxysuccinimide-ester/2712?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart