CH6371378488 Chem Impex International, Inc.

Nalpha-Boc-Nepsilon-Fmoc-L-lysine methyl ester

  • MDL number MFCD00236856
  • Cas Number : 133628-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101-108 °C
Purity 98
Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-boc-i-n-i-sup-epsilon-sup-fmoc-l-lysine-methyl-ester/2713?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-boc-i-n-i-sup-epsilon-sup-fmoc-l-lysine-methyl-ester/2713?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart