CH6371378489 Chem Impex International, Inc.

Nalpha-Boc-Nepsilon-trifluoroacetyl-L-lysine N-hydroxysuccinimide ester

  • MDL number MFCD00038649
  • Cas Number : 34695-46-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-126 °C
Purity 98
Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-boc-i-n-i-sup-epsilon-sup-trifluoroacetyl-l-lysine-i-n-i-hydroxysuccinimide-ester/2714?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-boc-i-n-i-sup-epsilon-sup-trifluoroacetyl-l-lysine-i-n-i-hydroxysuccinimide-ester/2714?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart