CH6371378497 Chem Impex International, Inc.

Na-Boc-Nd-Z-L-ornithine N-hydroxysuccinimide ester

  • MDL number MFCD00236870
  • Cas Number : 57225-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134-141
Synonym Boc-L-Orn(Z)-OSu
Purity 96
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-boc-i-n-i-sup-d-sup-z-l-ornithine-i-n-i-hydroxysuccinimide-ester/2721?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-boc-i-n-i-sup-d-sup-z-l-ornithine-i-n-i-hydroxysuccinimide-ester/2721?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart