CH6371378500 Chem Impex International, Inc.

Boc-D-phenylalanine N-hydrosuccinimide ester

  • MDL number MFCD00069688
  • Cas Number : 3674-18-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147-154
Synonym Boc-D-Phe-OSu
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-d-phenylalanine-i-n-i-hydrosuccinimide-ester/2725?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-d-phenylalanine-i-n-i-hydrosuccinimide-ester/2725?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart