CH6371378502 Chem Impex International, Inc.

Boc-L-phenylglycine N-hydroxysuccinimide ester

  • MDL number MFCD00190826
  • Cas Number : 201152-47-8
  • Molecular Weight : 348.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147 - 151 °C
Purity 98
Storage temp. Store at 0 - 8 °C
Physical Form White crystal powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-phenylglycine-i-n-i-hydroxysuccinimide-ester/2727?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-phenylglycine-i-n-i-hydroxysuccinimide-ester/2727?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart