CH6371378504 Chem Impex International, Inc.

Boc-O-benzyl-L-serine dicyclohexylammonium salt

  • MDL number MFCD00067670
  • Cas Number : 10342-01-5
  • Molecular Weight : 476.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133-139 °C
Purity 99
Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-o-i-benzyl-l-serine-dicyclohexylammonium-salt/2729?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-o-i-benzyl-l-serine-dicyclohexylammonium-salt/2729?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart