CH6371378505 Chem Impex International, Inc.

Boc-O-benzyl-L-serine 4-nitrophenyl ester

  • MDL number MFCD02094732
  • Cas Number : 16948-39-3
  • Molecular Weight : 416.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form Clear, yellow oil
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-o-i-benzyl-l-serine-4-nitrophenyl-ester/2730?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-o-i-benzyl-l-serine-4-nitrophenyl-ester/2730?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart