CH6371378506 Chem Impex International, Inc.

Boc-O-benzyl-D-serine N-hydroxysuccinimide ester

  • MDL number MFCD00236873
  • Cas Number : 82155-85-9
  • Molecular Weight : 392.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106-110 °C
Purity 98
Storage temp. Store at = -4 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-o-i-benzyl-d-serine-i-n-i-hydroxysuccinimide-ester/2731?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-o-i-benzyl-d-serine-i-n-i-hydroxysuccinimide-ester/2731?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart