CH6371378507 Chem Impex International, Inc.

Boc-L-threonine N-hydroxysuccinimide ester

  • MDL number MFCD00037904
  • Cas Number : 63076-44-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133-137 �C
Synonym Boc-L-Thr-OSu
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-threonine-i-n-i-hydroxysuccinimide-ester/2732?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-threonine-i-n-i-hydroxysuccinimide-ester/2732?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart