CH6371378509 Chem Impex International, Inc.

Na-Boc-Nin-formyl-L-tryptophan N-hydroxysuccinimide ester

  • MDL number MFCD00236877
  • Cas Number : 70601-13-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 153-160
Synonym Boc-L-Trp(For)-OSu
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-i-n-i-sup-in-sup-formyl-l-tryptophan-i-n-i-hydroxysuccinimide-ester/2734?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-i-n-i-sup-in-sup-formyl-l-tryptophan-i-n-i-hydroxysuccinimide-ester/2734?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart