CH6371378513 Chem Impex International, Inc.

Boc-L-tyrosine 4-nitrophenyl ester

  • MDL number MFCD00076991
  • Cas Number : 20866-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146-155
Synonym Boc-L-Tyr-ONp
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-tyrosine-4-nitrophenyl-ester/2738?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-tyrosine-4-nitrophenyl-ester/2738?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart